Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MAP1LC3C Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CQ7
75929-854EA 518 USD InStock
75929-854
Anti-MAP1LC3C Rabbit Polyclonal Antibody
Antibodies
Microtubule-associated proteins (MAPs) regulate microtubule stability and play critical roles in neuronal development and plasticity (1). MAP1LC3C belongs to the MAP1 LC3 family of proteins that form mature complexes with MAP1A and MAP1B which are thought to be important in the formation and development of axons and dendrites (2). MAP1LC3C is one of three isoforms of MAP1LC3, the mammalian homolog of yeast ATG8, an essential autophagy protein. These isoforms exhibit distinct expression patterns and MAP1LC3C, like MAP1LC3A but not MAP1LC3B, is post-translationally modified, suggesting the three isoforms may have different physiological functions (3).

Anti-MAP1LC3C Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 17 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MAP1LC3C
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty