Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MC4R Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CR7
75929-878EA 518 USD InStock
75929-878
Anti-MC4R Rabbit Polyclonal Antibody
Antibodies
The melanocortin-4 receptor (MC4R) is a member of the superfamily of seven transmembrane G-protein coupled receptors that are involved in multiple signal transduction pathways including the cAMP and MAPK signaling pathways. It is thought that the melanocortin system modulates energy expenditure and insulin sensitivity; activation of the MC4R results in the inhibition of c-Jun N-terminal kinase (JNK) activity and promotes insulin signaling. MC4R-null mice display maturity onset obesity characterized by hyperphagia, increased adiposity, hyperinsulinaemia and hyperleptinaemia, suggesting that like other obesity-linked genes such as FTO, PTER, and NPC1, MC4R is a potential candidate target for the treatment of obesity.

Anti-MC4R Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 37 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MC4R
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty