Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MCM7 Mouse Monoclonal Antibody [Clone: M3]

Supplier: Rockland Immunochemicals
200-301-P40
75929-888EA 528 USD InStock
75929-888
Anti-MCM7 Mouse Monoclonal Antibody [Clone: M3]
Antibodies
Initiation of DNA replication is a complex process involving the concerted action of many proteins. The chromosome 7-linked gene referred to by Nakatsuru et al. (1995) as MCM2 has been designated MCM7. The MCM2-7 complex is comprised of 6 subunits, MCM2 through MCM7, and is a ring-shaped heterohexameric ATPase involved in DNA replication. MCM7 is located in 7q21.3-q22.1. This antibody is suitable for researchers interested in cancer research and Cell Cycle research.

Anti-MCM7 is useful for Immunocytochemistry and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~81.3 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: MCM7
Clonality: Monoclonal
Clone: M3
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty