Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MECP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X39
75929-922EA 518 USD InStock
75929-922
Anti-MECP2 Rabbit Polyclonal Antibody
Antibodies
MeCP2 is a chromosomal protein with abundant binding sites in the chromatin. It belongs to the family of methyl CpG binding proteins which also comprises MBD1, MBD2, MBD3 and MBD4. MeCP2 can bind specifically to methylated promoters, thereby repressing transcription. This transcriptional repression is mediated through interaction with histone deacetylase and the corepressor SIN3A. MeCP2 also is essential for development. Mutations in MeCP2 are the cause of several types of mental retardation including Rett syndrome, a progressive neurological disorder that causes mental retardation in females and mental retardation syndromic X-linked type 13, and may also be involved in Angelman syndrome and susceptibility to some types of autism. Anti-MeCP2 Antibody is ideal for research in Epigenetics, Embryonic Development and Neuroscience.

Anti-MeCP2 Antibody is suitable for Chromatin Imunoprecipitation, ELISA and Western Blots. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 70 kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: MECP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty