Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MFSD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CU0
75929-930EA 518 USD InStock
75929-930
Anti-MFSD1 Rabbit Polyclonal Antibody
Antibodies
Multidrug transporters, such as MFSD1, are membrane proteins that expel a wide spectrum of cytotoxic compounds from the cell and render cells resistant to multiple drugs. The Major Facilitator Superfamily (MFS) is a large and diverse group of secondary transporters that include uniporters, symporters, and antiporters. Members of this family are found in all living organisms and are highly represented in bacteria. MFS members are capable of transporting various substrates such as sugars, polyols, drugs, neurotransmitters, amino acids, peptides, and inorganic anions, although most members are substrate-specific. MFS have provided important insight into the mechanism underlying multidrug transport. MFSD1 is still poorly understood.

Anti-MFSD1 Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 51 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MFSD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty