Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MMP16 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P76
75929-964EA 487 USD InStock
75929-964
Anti-MMP16 Rabbit Polyclonal Antibody
Antibodies
The matrix metalloproteinase 16(MMP16) protein consists of 604 amino acids and has a characteristic MMP domain structure, which gene is mapped on human chromosome 8q21. Additionally, MMP16 has a C-terminal extension containing a potential transmembrane domain, similar to MMP14 , MMP15 , and MMP17. Furthermore, it is membrane-bound and is a member of the membrane-type MMPs that are a subclass in the MMP family since the other members lack a C-terminal transmembrane domain and are secreted as soluble forms. MMP16 is expressed as a 12-kb transcript in brain, placenta, heart, and some carcinoma cell lines, but is not detectably expressed in lung, kidney, liver, spleen, and muscle. This antibody is suitable for researchers interested in cancer research.

Anti-MMP16 is useful for Immunohistochemistry -P, -F, and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~69.5 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: MMP16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty