Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Goat Polyclonal Antibody (DL800)

Supplier: Rockland Immunochemicals
617-145-012
75930-016EA 130 USD InStock
75930-016
Anti-IgG Goat Polyclonal Antibody (DL800)
Antibodies
Anti-Monkey IgG (gamma chain) antibody generated in goat detects specifically monkey IgG (gamma chain). This secondary antibody anti-Monkey is ideal for investigators who routinely perform ELISA, Sandwich ELISA, titration assays, western-blot, immunoprecipitation and more generally immunoassays.

Anti-Monkey IgG (gamma chain) is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays. The emission spectra for this DyLight™ conjugate match the principle output wavelengths of most common fluorescence instrumentation. Specific conditions for reactivity and signal detection should be optimized by the end user.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: DL800
Epitope:
Host: Goat
Isotype:
Reactivity: Monkey

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty