Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MRGPRX1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-P80
75930-142EA 487 USD InStock
75930-142
Anti-MRGPRX1 Rabbit Polyclonal Antibody
Antibodies
MAS-related G-protein coupled receptor, member C, also known as MRGPRC or MRGPRX1, is a human gene. The Mas-related genes (Mrgs) comprise a family of >50 G protein-coupled receptors (GPCRs), many of which are expressed in specific subsets of nociceptive sensory neurons in mice. In contrast, humans contain a related but nonorthologous family of genes, called MrgXs or sensory neuron-specific receptors, of which many fewer appear to be expressed in sensory neurons. This antibody is suitable for researchers interested in neuroscience research.

Anti-MRGPRC is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~37.1 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: MRGPRX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty