Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-STXBP2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X16
75930-160EA 518 USD InStock
75930-160
Anti-STXBP2 Rabbit Polyclonal Antibody
Antibodies
Munc-18 ( mammalian homologue of Unc-18) is a protein that is thought to be involved in regulating exocytosis due, at least in part, to it ability to bind syntaxin. Munc18-1 is a neuron-specific member of the Sec1/Munc18 protein family that binds to syntaxin1A and is thought to stabilize the complex. The function of Munc-18 is thought to be regulated by PKC phosphorylation of Ser515 on the Munc-18 protein. Munc-18 pS515 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser515 Munc18 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and Immunohistochemistry, and it is specific for the ~65 kDa Munc-18 protein phosphorylated at Ser515. Immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: STXBP2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty