Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MYH8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CW8
75930-170EA 518 USD InStock
75930-170
Anti-MYH8 Rabbit Polyclonal Antibody
Antibodies
Myosins are actin-based motor proteins that function in the generation of mechanical force in eukaryotic cells (1). MYH8 (myosin, heavy chain 8, skeletal muscle, perinatal) is a member of the class II or conventional myosin heavy chains, and functions in skeletal muscle contraction (2,3). This gene is predominantly expressed in fetal skeletal muscle. MYH8 is regulated by phosphorylation via myosin light chain kinase (MLCK) and by intracellular Ca2+ concentrations (3). A mutation in this gene results in trismus-pseudocamptodactyly syndrome (4).

Anti-MYH8 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 223 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: MYH8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty