Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MB Mouse Monoclonal Antibody [Clone: JAN-12]

Supplier: Rockland Immunochemicals
200-301-P42
75930-172EA 528 USD InStock
75930-172
Anti-MB Mouse Monoclonal Antibody [Clone: JAN-12]
Antibodies
Human myoglobin has 152 residues. Two myoglobin variants were found. The myoglobin locus mapped to 22q11-22q13. The myoglobin gene is about 10.5 kb long and contains two introns as in the case with hemoglobin genes. Myoglobin may serve a variety of functions in muscular oxygen supply, such as O(2) storage, facilitated O(2) diffusion, and myoglobin-mediated oxidative phosphorylation. This antibody is suitable for researchers interested in cancer research.

Anti-Myoglobin is useful for Immuohistochemistry -P. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~17.2 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: MB
Clonality: Monoclonal
Clone: 43112
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty