Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NAM-DH Goat Polyclonal Antibody (BAC)

Supplier: Rockland Immunochemicals
200-106-228S
75930-206EA 148 USD InStock
75930-206
Anti-NAM-DH Goat Polyclonal Antibody (BAC)
Antibodies
Anti-N-Acylmannosamine-1-Dehydrogenase recognizes the protein N-Acylmannosamine 1-Dehydrogenase, a member of the oxidoreductase family that act on CH-OH donors and NAD+ or NADP+ as acceptors. N-acyl-D-mannosamine 1-dehydrogenase catalyzes the reaction in which N-acyl-D-mannosamine and NAD+ are converted to N-acyl-D-mannosaminolactone, NADH, and H+.

Anti-N-Acylmannosamine-1-Dehydrogenase antibody has been assayed against 1.0 µg of N-Acylmannoseamine-1-Dehydrogenase in a standard capture ELISA using Peroxidase Conjugated Streptavidin #S000-03 and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid]) code # ABTS-100 as a substrate for 30 minutes at room temperature.  A working dilution of 1:7,000 to 1:35,000 of the reconstitution concentration is suggested for this product.

Type: Primary
Antigen: NAM-DH
Clonality: Polyclonal
Clone:
Conjugation: BAC
Epitope:
Host: Goat
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty