Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NGFRAP1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
200-401-CX2
75930-208EA 518 USD InStock
75930-208
Anti-NGFRAP1 Rabbit Polyclonal Antibody
Antibodies
The p75 neurotrophin receptor (p75NTR) is a member of the tumor necrosis receptor superfamily and can mediate cell death and cell survival in response to nerve growth factor (NGF). The p75NTR-associated cell death executor (NADE) mediates apoptosis by interacting with the cell death domain of p75NTR following the binding of NGF by p75NTR. Recent studies have shown that NADE also interacts with second mitochondria-derived activator of caspase (Smac). Co-expression of NADE and Smac promotes TRAIL-induced apoptosis and inhibits XIAP-mediated Smac ubiquitization. It has been suggested that it is this interaction between NADE and Smac that allows apoptosis to proceed. Finally, although initially discovered as an mRNA expressed in ovarian granulosa cells, NADE has been suggested to play a role in the neuronal death seen in epileptic brain damage.

Anti-NADE Antibody has been tested for use in ELISA, Western Blotting, and Immunohistochemistry. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 13 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NGFRAP1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty