Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NLRP3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H02
75930-228EA 518 USD InStock
75930-228
Anti-NLRP3 Rabbit Polyclonal Antibody
Antibodies
NALP3 antibody detects human NALP3. NALP3, a member of Nod-like receptors, has a crucial role in inflammation and immunity and may be a proximal sensor of cellular stress and danger signals. NALP3 forms a caspase-1 activating molecular complex termed the inflammasome. The inflammasome allows activation of IL-1beta, the key player of the inflammation and fever. NALP3 gene encodes a pyrin like protein which contains a pyrin domain, a nucleotide binding site (NBS) domain, and a leucine rich repeat (LRR) motif. NALP3 protein interacts with apoptosis associated speck like protein containing a CARD and may function as an inducer of apoptosis and an activator of NF-kappaB signaling. Defects in NALP3 have been associated with familial cold autoinflammatory syndrome (FCAS), Muckle-Wells syndrome (MWS), chronic infantile neurological cutaneous and articular (CINCA) syndrome, and neonatal-onset multisystem inflammatory disease (NOMID). Anti-NALP3 antibodies are ideal for investigators involved in cytokine and growth factor research.

Anti-NALP3 Antibody is suitable for use in E,WB, and IF. Expect a band approximately ~118.1kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: NLRP3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty