Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NANOS1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CY8
75930-236EA 518 USD InStock
75930-236
Anti-NANOS1 Rabbit Polyclonal Antibody
Antibodies
Nanos1 is one of three known mammalian homologs to the Drosophila gene nanos. Nanos1 is an RNA-binding protein containing a zinc-finger motif and is expressed in the developing nervous system and continues in the adult brain. Interestingly, unlike mice deficient in either nanos2 or nanos3, mice lacking the nanos1 gene develop normally with no sign of abnormalities. Recently it has been found that expression of nanos1 mRNA is down-regulated by E-cadherin in a human breast cancer cell line and the amino-terminal domain on Nanos1 interacts with the E-cadherin-binding protein p120ctn. Furthermore, overexpression of Nanos1 in human colorectal DLD1 cancer cells functionally abolished cell-cell adhesion, allowing the cancer cells to develop strong migratory and invasive properties. These results suggest that targeting Nanos1 might prove an effective strategy in the treatment of E-cadherin-negative tumors.

Anti-Nanos1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 30 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NANOS1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty