Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NDST2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-CZ6
75930-258EA 518 USD InStock
75930-258
Anti-NDST2 Rabbit Polyclonal Antibody
Antibodies
The N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 2 (NDST2) protein, also known as N-heparan sulfate sulfotransferase 2, and the highly related protein NDST1 have key roles in the sulfation of heparan sulfate proteoglycans, molecules that are present of cell surfaces and the extracellular matrix (1,2). NDST1 and NDST2 are dispensable for mesodermal differentiation into osteoblasts but necessary for induction of adipocytes and neural cells (3).

Anti-NDST2 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 101 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NDST2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty