Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neuroligin-1 Mouse Monoclonal Antibody [Clone: S97A-31]

Supplier: Rockland Immunochemicals
200-301-W75
75930-310EA 540 USD InStock
75930-310
Anti-Neuroligin-1 Mouse Monoclonal Antibody [Clone: S97A-31]
Antibodies
Neuroligin-1 is a neuronal cell surface protein belonging to the type-B carboxylesterase/lipase family. It is a necessary component in the maturation of excitatory synapses for their normal, functional development, and is necessary to the regulation of synaptic plasticity and the development of long-term memory within the adult amygdala in mammals. It is believed to participate in cell-cell-interaction through the assembly of intracellular junction by the binding of beta-neurexins, and may also be a factor in the maintenance and assembly of synaptic junctions. It is also thought to have involvement in excitatory synaptic specification. Within brain tissue, Neuroligin-1 is primarily observed in neurons and spinal cord. Anti-Neuroligin-1 is ideal for research in Neuroscience, including narcolepsy and synaptic neurotransmission: GABAergic inhibition, as well as Cell Adhesion.

Anti-Neuroligin 1 Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~120kDa on specific lysates or tissues. Does not cross react with other Neuroligins. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Neuroligin-1
Clonality: Monoclonal
Clone: S97A-31
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty