Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Neuroligin-3 Mouse Monoclonal Antibody (RPE) [Clone: S110-29]

Supplier: Rockland Immunochemicals
200-308-W83
75930-380EA 539 USD InStock
75930-380
Anti-Neuroligin-3 Mouse Monoclonal Antibody (RPE) [Clone: S110-29]
Antibodies
Neuroligins are Type I membrane proteins enriched in synaptic plasma membranes and clustered in synaptic clefts and postsynaptic densities. They have been characterized as neuronal cell surface proteins and are thought to be involved in cell-cell-interactions by forming intercellular junctions through binding to beta-neurexins. They play a major role in the formation or maintenance of synaptic junctions. They are also thought to be involved in the specification of excitatory synapses. Neuroligins interact with beta-neurexins and this interaction is involved in the formation of functional synapses. Anti-Neuroligin-3 is ideal for research in Neuroscience, including autism research, and Cell Adhesion and Communication.

Anti-Neuroligin-3 R-PE Conjugated Antibody is suitable for Western Blots, Immunohistochemistry and Immunocytochemistry. Expect a band approximately ~110kDa on specific lysates or tissues. Does not cross react with Neuroligin -1, -2, or -4. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Neuroligin-3
Clonality: Monoclonal
Clone: S110-29
Conjugation: RPE
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty