Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NBN Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DA9
75930-426EA 518 USD InStock
75930-426
Anti-NBN Rabbit Polyclonal Antibody
Antibodies
NIBRIN (NBN) is a member of the double-strand break repair complex MRE11/RAD50/NBN (MRN) which is involved in DNA double-strand break repair, DNA damage-induced checkpoint activation and plays a critical role in the maintenance of chromosome integrity. NIBRIN contains two modules found in cell cycle checkpoint proteins, a forkhead-associated domain adjacent to a breast cancer carboxy-terminal domain. Mutations in this gene are associated with Nijmegen breakage syndrome and maybe the cause of cancer predisposition and aplastic anemia.

Anti-NIBRIN Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 85 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NBN
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty