Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKX2-1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DC2
75930-458EA 518 USD InStock
75930-458
Anti-NKX2-1 Rabbit Polyclonal Antibody
Antibodies
NKX2-1 (NK2 homeobox 1) has been identified as a thyroid-specific transcription factor that binds and activates the promoter of thyroid specific genes such as thyroglobulin and thyrotropin receptor. NKX2-1 is crucial in the maintenance of the thyroid differentiation phenotype and morphogenesis. It contains one homeobox DNA-binding domain. NKX2-1 also may play a role in lung development and surfactant homeostasis. Mutations and deletions in this gene are associated with benign hereditary chorea, congenital hypothyroidism, neonatal respiratory distress, and may be associated with thyroid cancer. NKX2-1 has been suggested to be a candidate suppressor of malignant progression.

Anti-NKX2-1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 39 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NKX2-1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty