Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NKX2-8 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DC6
75930-466EA 518 USD InStock
75930-466
Anti-NKX2-8 Rabbit Polyclonal Antibody
Antibodies
NKX2-8 (NK2 homeobox 8) is a member of a family of transcription factors that are involved in embryonic development and cell fate. It is expressed in the ventral foregut, the developing heart, the epithelial layers of the branchial arches and in the dorsal mesoderm. In conjunction with related protein, NKX2-5, NKX2-8 may play a role in cardiac embryonic development. NKX2-8 is also thought to be involved in lung development and is suspected of being an oncogene in lung cancer that is activated by way of gene amplification at chromosome 14q13.

Anti-NKX2-8 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 26 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NKX2-8
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty