Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-GRIN2B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X17
75930-472EA 518 USD InStock
75930-472
Anti-GRIN2B Rabbit Polyclonal Antibody
Antibodies
The NMDA receptor (NMDAR) plays an essential role in memory, neuronal development and it has also been implicated in several disorders of the central nervous system including Alzheimer’s, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B-subunit of the NMDA Receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Phosphorylation of Ser1166 is thought to play an essential role in memory and neuronal development. NMDA pS1166 Recepor NR2B Subunit Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Ser1166 NMDA Receptor NR2B Subunit Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~180 kDa NMDAR NR2B subunit phosphorylated at Ser1166. Immunolabeling of the NMDA NR2B subunit band is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: GRIN2B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty