Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NLRX1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DD1
75930-484EA 518 USD InStock
75930-484
Anti-NLRX1 Rabbit Polyclonal Antibody
Antibodies
NOD5, also known as NLRX1, is a member of the NOD (nucleotide-binding oligomerization domain) family, a group of proteins that are involved in innate immune defense. NOD5 localizes to the mitochondrial outer membrane and interacts with the virus-induced signaling adapter protein VISA. Unlike a subset of NOD-like receptors (NLRs) such as NOD1 and NOD2 which trigger pro-inflammatory cascades, and other NLRs that induce the caspase 1 inflammasome in response to immune challenges, NOD5 amplifies NF-kB and JNK pathways by inducing reactive oxygen species production.

Anti-NOD5 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 108 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NLRX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty