Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOL3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DD4
75930-490EA 518 USD InStock
75930-490
Anti-NOL3 Rabbit Polyclonal Antibody
Antibodies
Apoptosis, also known as programmed cell death, plays major roles in development and normal tissue turnover in addition to tumor formation. Apoptosis is regulated by death domain (DD) and/or caspase recruitment domain (CARD) containing molecules and the caspase family of proteases. CARD domain containing cell death regulators include RAIDD, Apaf-1, caspase-9, and caspase-2. A novel CARD domain containing protein was recently identified and designated ARC for apoptosis repressor with CARD. An alternate splicing isoform of ARC was identified as Nop30. While ARC interacts with caspase-2 and -8 and suppresses apoptosis induced by cell death adapters FADD and TRADD and by cell death receptors Fas, TNFR-1 and DR3, Nop30 multimerizes and binds to the splicing factor SRp30c and may act to influence alternative splice site selection in vivo. The Nop30 antibody will not detect ARC protein.

Anti-Nop30 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 24 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NOL3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty