Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CYBB Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-R16
75930-516EA 487 USD InStock
75930-516
Anti-CYBB Rabbit Polyclonal Antibody
Antibodies
NOX2(NADPH OXIDASE 2), also called CYBB(CYTOCHROME b(-245), BETA SUBUNIT), p91-PHOX or GP91-1, is a human gene encoding a glycoprotein.NOX2 is an essential component of phagocytic NADPH-oxidase, a membrane-bound enzyme complex that generates large quantities of microbicidal superoxide and other oxidants upon activation. It is mapped on Xp11.4. NOX2 is a heterodimer composed of an alpha chain of relative molecular mass 23 kD and a beta chain of 76 to 82 kD. NOX2 assembled on DC phagosomes in a gp91-phox subunit-dependent manner, and that reactive oxygen species were produced in a more sustained manner in immature DC phagosomes than in macrophage phagosomes .As a major player in innate immune responses in neutrophils,NOX2 is also involved in adaptive immunity through its activity in DCs. In heart cells, physiologic stretch rapidly activates reduced-form NOX2 to produce reactive oxygen species (ROS) in a process dependent on microtubules (X-ROS signaling). This antibody is suitable for researchers interested in cancer research.

Anti-NOX2 is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~65.3 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: CYBB
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty