Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NOX4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DE1
75930-520EA 518 USD InStock
75930-520
Anti-NOX4 Rabbit Polyclonal Antibody
Antibodies
The NOX family of NAPDH oxidases is comprised of seven transmembrane proteins that oxidize intracellular NAPDH/NADH, causing electron transport across the membrane and the reduction of molecular oxygen to superoxide (1). NOX4 is is expressed in multiple tissues and catalyzes the reduction of molecular oxygen to various reactive oxygen species (ROS) (2,3). Unlike other NOX proteins, NOX4 does not require cytosolic subunits and thus is constitutively active (4). The function of NOX4 remains unclear as it plays both protective and deleterious roles in cellular metabolism.

Anti-NOX4 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 67 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: NOX4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty