Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-NrCAM Mouse Monoclonal Antibody (ALP) [Clone: S364-51]

Supplier: Rockland Immunochemicals
200-305-W74
75930-540EA 539 USD InStock
75930-540
Anti-NrCAM Mouse Monoclonal Antibody (ALP) [Clone: S364-51]
Antibodies
Neuronal cell adhesion molecule (NrCAM) is a cell surface protein of the immunoglobulin (Ig) superfamily. NrCAM (also known as Bravo) contains six Ig domains, five fibronectin repeats, a transmembrane region and an intracellular domain. NrCAM is expressed in brain, spinal cord, peripheral nervous system and pancreas. In the spinal cord, NrCAM acts as a ligand for axonin-1 to guide commissural axons across the floor plate. NrCAM also acts as a ligand for F3 to control actin-dependent growth cone motility. NrCAM interacts with neurofascin and may facilitate the clustering of the cystoskeletal protein ankyrin G and the voltage-dependent sodium channel proteins at the node of Ranvier. NrCAM expression may play a role in the severity of certain types of tumors. NrCAM is overexpressed in high-grade astrocytomas, gliomas and glioblastoma tumor tissues. Anti-NrCam is ideal for research in Cancer and Neuroscience.
In the pancreas, NrCAM expression is upregulated in intraductal hyperplasia. Antisense NrCAM reduces the tumorigenic properties of human glioblastoma cells in vitro and slowed tumor growth in vivo. The gene encoding human NrCAM maps to chromosome 7q31.1-q31.2.

Anti-NrCAM Alkaline Phosphatase Conjugated Antibody is suitable for Western Blots and Immunocytochemistry. Expect a band approximately ~160kDa on specific lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: NrCAM
Clonality: Monoclonal
Clone: S364-51
Conjugation: ALP
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty