Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-ORC2 Mouse Monoclonal Antibody

Supplier: Rockland Immunochemicals
200-301-V05
75930-652EA 528 USD InStock
75930-652
Anti-ORC2 Mouse Monoclonal Antibody
Antibodies
ORC2 is one of the six subunits of the ORC (origin recognition complex) that binds origins of replication. The binding of ORC to DNA is ATP-dependent, but so far, specific DNA sequences that define origins of replication have not been identified. The ORC is required to assemble the pre-replication complex necessary to initiate DNA replication. The ORC is regulated in a cell cycle-dependent manner. It is assembled at the end of the anaphase of mitosis and disassembled when cells enter the S phase. Anti-ORC2 Antibody is ideal for research in DNA Replication, Cell Cycle Control and Cancer.

Anti-ORC2 Antibody is suitable for Western Blots. Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 66kDa in the appropriate cell lysate or extract.

Type: Primary
Antigen: ORC2
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty