Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-CDKN1B Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-K08
75930-680EA 518 USD InStock
75930-680
Anti-CDKN1B Rabbit Polyclonal Antibody
Antibodies
p27 kip1 pS140 is a cyclin-dependent kinase inhibitor that shares a limited similarity with CDK inhibitor CDKN1A/p21.  p27 binds to and prevents the activation of cyclin E-CDK2 or cyclin D-CDK4 complexes, and thus controls the cell cycle progression at G1. The degradation of this protein, which is triggered by its CDK dependent phosphorylation and subsequent ubiquitination by SCF complexes, is required for the cellular transition from quiescence to the proliferative state. Phosphorylation occurs on serine, threonine and tyrosine residues. Anti-p27 kip1 pS140 antibody is ideal for researchers interested in cell cycle and cancer research.

Anti-p27kip1 pS140 antibody is suitable for use in ELISA, immunoprecipitation, co-immunoprecipitation and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at ~27 kDa in size corresponding to p27 by western blotting in the appropriate cell lysate or extract. Anti-p27 is suitable for the detection by immunoblot of human p27. Optimal titers for other applications should be determined by the researcher.

Type: Primary
Antigen: CDKN1B
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty