Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-SQSTM1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X19
75930-718EA 518 USD InStock
75930-718
Anti-SQSTM1 Rabbit Polyclonal Antibody
Antibodies
p62, also known as sequestosome1 (SQSTM1), is a shuttle protein transporting polyubiquitinated proteins for both proteasomal and lysosomal degradation. p62 is an integral component of inclusions in brains of various neurodegenerative disorders, including Alzheimer disease (AD) neurofibrillary tangles (NFTs) and Lewy bodies in Parkinson disease. p62 plays an important role in the protection of cells from the toxicity of misfolded proteins by enhancing aggregate formation especially in the later stages. Phosphorylation of Ser28 has recently been demonstrated to be related to the pathogenesis of Parkinson’s disease. p62 pT269/pS272 Antibody is ideal for researchers interested in neuroscience research.

Anti-Phospho-Thr269/Ser272 p62 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~62 kDa p62 protein phosphorylated at Thr269 and Ser272. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: SQSTM1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty