Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PARK2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DJ9
75930-730EA 518 USD InStock
75930-730
Anti-PARK2 Rabbit Polyclonal Antibody
Antibodies
Parkinson’s disease (PD) is a neurodegenerative disease whose symptoms include tremors, rigidity, bradykinesia, and postural instability (1). Mutations in the PARK2 gene are known to cause Parkinson disease and autosomal recessive juvenile Parkinson disease (2). The PARK2 protein is a component of a multiprotein E3 ubiquitin ligase complex that mediates the targeting of substrate proteins for proteasomal degradation (3). Recent studies have suggested that PARK2 expression reduces the mitochondrial accumulation of the apoptosis protein Bax under basal conditions and directly ubiquitinates Bax, thereby promoting cell survival (4).

Anti-PARK2 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 52 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PARK2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty