Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PXN Mouse Monoclonal Antibody [Clone: PAX-14]

Supplier: Rockland Immunochemicals
200-301-P44
75930-740EA 528 USD InStock
75930-740
Anti-PXN Mouse Monoclonal Antibody [Clone: PAX-14]
Antibodies
The paxillin gene can be alternatively spliced to include 1 of 2 alternative exons, generating beta and gamma isoforms. Paxillin is a 68-kDa focal adhesion protein that is phosphorylated on tyrosine residues in fibroblasts in response to transformation by v-src, treatment with platelet-derived growth factor, or cross-linking of integrins. The 68-kD protein (paxillin) is a cytoskeletal component that localizes to the focal adhesions at the ends of actin stress fibers in chicken embryo fibroblasts. It is also present in the focal adhesions of Madin-Darby bovine kidney (MDBK) epithelial cells but is absent, like talin, from the cell-cell adherens junctions of these cells. This antibody is suitable for researchers interested in growth factors and signal trasduction.

Anti-Paxillin is useful for Immunohistochemistry and Western Blot. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~64.5 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: PXN
Clonality: Monoclonal
Clone: PAX-14
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty