Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PDCD1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DK2
75930-760EA 518 USD InStock
75930-760
Anti-PDCD1 Rabbit Polyclonal Antibody
Antibodies
Cell-mediated immune responses are initiated by T lymphocytes that are themselves stimulated by cognate peptides bound to MHC molecules on antig en-presenting cells (APC). T-cell activation is generally self-limited as activated T cells express receptors such as PD-1 (also known as PDCD-1) that mediate inhibitory signals from the APC. PD-1 can bind two different but related ligands, PDL-1 and PDL-2. Upon binding to either of these ligands, signals generated by PD-1 inhibit the activation of the immune response in the absence of "danger signals" such as LPS or other molecules associated with bacteria or other pathogens. Evidence for this is seen in PD1-null mice who exhibit hyperactivated immune systems and autoimmune diseases.

Anti-PD-1 Antibody has been tested for use in ELISA, Western Blotting, Immunofluorescence, and Immunohistochemistry. Expect a band at approximately 32 kDa in Western Blots using positive control THP-1 whole cell lysate, Human brain tissue for IHC, or other specific cell lysates and tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: PDCD1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty