Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PHF14 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DM1
75930-866EA 518 USD InStock
75930-866
Anti-PHF14 Rabbit Polyclonal Antibody
Antibodies
PHF14, a novel plant homology domain (PHD) transcription factor, controls the proliferation of mesenchymal cells by regulating PDGFR alpha expression in embryonic development (1). PHD finger transcription factors are concerned with transcriptional regulation by interacting with modified histones (2), and dysregulation of several genes encoding PHD finger transcription factors, such as ING1 and RBP2, has been found in human cancer diseases suggesting these factors have an important role for cancer cell proliferation. PHF14 is a potential target for new treatments of lung fibrosis (1, 3).

Anti-PHF14 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 100 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PHF14
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty