Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIK3C3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DN6
75930-994EA 518 USD InStock
75930-994
Anti-PIK3C3 Rabbit Polyclonal Antibody
Antibodies
PI 3-kinase p100 (phosphoinositide-3-kinase p100 subunit), also known as hVps34 or PIK3C3 (phosphoinositide-3-kinase class III), is a member of the PI3/PI4-kinase family (1). It is a catalytic subunit of the PI3K complex that mediates formation of phosphatidylinositol 3-phosphate and ubiquitously expressed with a highest expression in skeletal muscle (1,2). PIK3C3 is involved in the endosome to lysosome transport and plays important roles in intracellular membrane trafficking and autophagy (3-5).

Anti-PIK3C3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 101 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PIK3C3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty