Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIK3R2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-R17
75930-996EA 487 USD InStock
75930-996
Anti-PIK3R2 Rabbit Polyclonal Antibody
Antibodies
PIK3R2(Phosphatidylinositol 3-kinase, regulatory subunit 2), also called p85-Beta, is an enzyme that in humans is encoded by the PIK3R2 gene. The PIK3R2 gene is mapped on 19p13.11. The NS1 protein from various influenza strains also bound p85-beta, but not p85-alpha. Binding of p85-beta and activation of PI3K required tyr89 of NS1, and mutant viruses expressing NS1 with a tyr89-to-phe substitution grew more slowly in cell culture than wildtype viruses. Using mouse embryonic fibroblasts, Park et al. showed that, in addition to regulating PI3K function, p85-alpha and p85-beta regulated the function of Xbp1s, a transcription factor that orchestrates the unfolded protein response (UPR) following endoplasmic reticulum (ER) stress. Overexpression of PIK3R2 in livers of ob/ob mice increased glucose tolerance and reduced blood glucose concentrations. This antibody is suitable for researchers interested in cancer research.

Anti-PIK3R2 is useful for Immunohistochemistry -P and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately ~81.5 kDa corresponding to the appropriate cell lysate or extract.

Type: Primary
Antigen: PIK3R2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty