Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PINK1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DN9
75931-004EA 518 USD InStock
75931-004
Anti-PINK1 Rabbit Polyclonal Antibody
Antibodies
The PTEN-induced putative kinase 1 (PINK1) is a serine/threonine protein kinase that localizes to mitochondria and is thought to protect cells from stress-induced mitochondrial dysfunction (reviewed in 1). PINK1 recruits the E3 ubiquitin ligase Parkin to mitochondria to initiate mitophagy, an autophagic process that clears damaged mitochondria within a cell (2). PINK1 is cleaved by the mitochondrial protease PARL (3). Mutations in this gene cause one form of autosomal recessive early-onset Parkinson disease (4).

Anti-PINK1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 63 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PINK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty