Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PIWIL1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DP2
75931-030EA 518 USD InStock
75931-030
Anti-PIWIL1 Rabbit Polyclonal Antibody
Antibodies
PIWI-L1 is a member of the PIWI subfamily of Argonaute proteins, evolutionarily conserved proteins containing both PAZ and Piwi motifs that are crucial for the biogenesis and function of small non-coding RNAs and play important roles in stem cell self-renewal, RNA silencing, and translational regulation in diverse organisms. PIWI-L1 is thought to play a role as an intrinsic regulator of the self-renewal capacity of germline and hematopoietic stem cells as removing PIWI-L1 protein from single germline stem cells significantly decreases the rate of their division. Recent studies have shown that overexpression of PIWI-L1 (as well as other PIWI-like proteins) are potential biomarkers for colon and other cancers.

Anti-PIWI-L1 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 99 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PIWIL1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty