Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PJA1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DP6
75931-038EA 518 USD InStock
75931-038
Anti-PJA1 Rabbit Polyclonal Antibody
Antibodies
Ubiquitinization is an important cellular degradation process requiring sequential reactions that are mediated by three enzymes: E1, E2 and E3. PJA1, also known as Praja1 and RING finger protein 70, is a 643 amino acid E2-dependent E3-ubiquitin ligase that is abundantly expressed in the brain (1,2). Through interaction and activation with the E2-ubiquitin ligase UBC4, PJA1 mediates substrate-specific ubiquitination via its RING finger domain and facilitates ubiquitination (3). Overexpression of PJA1 in gastrointestinal cancers suggests that it may be responsible for the degradation of some anti-oncogenic proteins (4,5).

Anti-PJA1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 71 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PJA1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty