Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PLAC2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DQ1
75931-048EA 518 USD InStock
75931-048
Anti-PLAC2 Rabbit Polyclonal Antibody
Antibodies
PLAC2 (placenta-specific protein 2) is a 151 amino acid protein that is encoded by a gene located on human chromosome 19p13. Based on its amino acid sequence, PLAC2 is predicted to be a multi-pass membrane protein. There are no reported studies or functions reported for this protein.

Anti-PLAC2 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 16 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PLAC2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty