Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-POU3F2 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DR4
75931-116EA 518 USD InStock
75931-116
Anti-POU3F2 Rabbit Polyclonal Antibody
Antibodies
POU3F2, also commonly known as brain-2, is a member of a family of POU domain genes expressed in mouse brain and is thought to be involved in the development of the neocortex and establishment of neural cell lineage. Recent studies suggest that POU3F2 may be involved with the development of neurodegenerative diseases as well as tumor development and proliferation. Along with the neural-lineage-specific transcription factors ASCL1 and MYT1L, POU3F2 can convert fibroblasts to functional neurons in vitro, a form of artificial stem cells termed induced neuronal (iN) cells, suggesting that these cells may be useful in the treatment of neurodegenerative diseases.

Anti-POU3F2 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 47 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: POU3F2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty