Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPFIA3 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DS2
75931-140EA 518 USD InStock
75931-140
Anti-PPFIA3 Rabbit Polyclonal Antibody
Antibodies
The PPFIA3 protein is a member of the LAR protein-tyrosine phosphatase-interacting protein (liprin) family, a family of transmembrane protein tyrosine phosphatases known to be important for axon guidance and mammary gland development (1). Similar to the other liprin family members, PPFIA3 can interact with ERC2, a member of the ELKS-Rab6-interacting protein-CAST family of proteins at the presynaptic termini, suggesting that these proteins may play a role in the molecular organization of presynaptic zones (2).

Anti-PPFIA3 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 133 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PPFIA3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty