Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PPM1D Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DS3
75931-142EA 518 USD InStock
75931-142
Anti-PPM1D Rabbit Polyclonal Antibody
Antibodies
Wild-type p53 induced phosphatase 1 (WIP1) / protein phosphatase magnesium-dependent 1 delta (PPM1D), a protein identified in the p53 DNA response pathway, is a member of the PP2C family of serine/threonine protein phosphatases which selectively inactivates p38 MAPK and dephosphorylates the ATM/ATR targets, Chk1 and p53 (1,2). Studies have shown that it is overexpressed in human cancers and is involved in the regulation of multiple DNA damage signaling pathways (2,3). PPM1D deletion results in a reduction of T and B cell function and compromised cell division, rendering cells resistant to becoming cancerous and slowing tumor development (4,5).

Anti-PPM1D Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 47 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PPM1D
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty