Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRDM16 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-H12
75931-146EA 518 USD InStock
75931-146
Anti-PRDM16 Rabbit Polyclonal Antibody
Antibodies
PRDM16 antibody detects human PRDM16. PRDM16 is a zinc finger transcription factor and contains an N-terminal PR domain. The reciprocal translocation t(1;3)(p36;q21) occurs in a subset of myelodysplastic syndrome (MDS) and acute myeloid leukemia (AML). This gene is located near the 1p36.3 breakpoint and has been shown to be specifically expressed in the t(1:3)(p36, q21)-positive MDS/AML. The translocation results in the overexpression of a truncated version of this protein that lacks the PR domain, which may play an important role in the pathogenesis of MDS and AML. Recent studies have shown that PRDM16 normally acts as a Smad3 binding protein that may be important for the development of orofacial structures through modulation of the TGF-beta signaling pathway. Other experiments have indicated that PRDM16 controls a bidirectional cell fate switch between skeletal myoblasts and brown fat cells. Anti-PRDM16 antibodies are ideal for investigators involved in enzyme and cytokines and growth factor research.

Anti-PRDM16 Antibody is suitable for use in ELISA, Western Blot, and Immunohistochemistry. Expect a band approximately ~140.2 kDa in western blot using positive control K562 whole cell lysates, or other specific cell lysates or tissues. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: PRDM16
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty