Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRICKLE1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DS8
75931-154EA 518 USD InStock
75931-154
Anti-PRICKLE1 Rabbit Polyclonal Antibody
Antibodies
PRICKLE1, also known as RILP or EPM1B, is a Disheveled-associated protein that serves as a nuclear translocation receptor for REST/NRSF and REST4 (1, 2). It contains three N-terminal LIM domains and three C-terminal nuclear localization signals. It localizes to the cytoplasm, as well as to the nuclear membrane and expressed at higher levels in placenta. PRICKLE1 is a negative regulator of the Wnt / beta-catenin signaling pathway and is a putative tumor suppressor in human HCCs. Defects in the gene encoding PRICKLE1 are associated with autosomal recessive progressive myoclonic epilepsy.

Anti-PRICKLE1 Antibody has been tested for use in ELISA, Western Blotting and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 94 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PRICKLE1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty