Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRICKLE4 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DT0
75931-158EA 518 USD InStock
75931-158
Anti-PRICKLE4 Rabbit Polyclonal Antibody
Antibodies
PRICKLE4, also known as over-expressed breast tumor protein (OBTP), is a member of the LIM family of proteins. It possesses two LIM domains as well as a PET domain and is thought to localize to the nucleus. Its expression in multiple malignant tissue differentiations indicate that it may play a role in cancer differentiation. Two other members of the LIM family, PRICKLE1 and PRICKLE2, are thought to function in the noncanonical WNT signaling pathway which regulates intracellular calcium release and planar cell polarity, suggesting the PRICKLE4 may play a similar role.

Anti-PRICKLE4 Antibody has been tested for use in ELISA, Western Blotting, Immunocytochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 38 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PRICKLE4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty