Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Protocadherin-Gamma-A1 Mouse Monoclonal Antibody (BAC) [Clone: S159-5]

Supplier: Rockland Immunochemicals
200-306-W68
75931-220EA 539 USD InStock
75931-220
Anti-Protocadherin-Gamma-A1 Mouse Monoclonal Antibody (BAC) [Clone: S159-5]
Antibodies
The protocadherin gamma gene cluster is one of three related clusters tandemly linked on chromosome five. These gene clusters have an immunoglobulin-like organization, suggesting that a novel mechanism may be involved in their regulation and expression. The gamma gene cluster includes 22 genes divided into 3 subfamilies. Subfamily A contains 12 genes, subfamily B contains 7 genes and 2 pseudogenes, and the more distantly related subfamily C contains 3 genes.
The tandem array of 22 large, variable region exons are followed by a constant region, containing 3 exons shared by all genes in the cluster. Each variable region exon encodes the extracellular region, which includes 6 cadherin ectodomains and a transmembrane region. The constant region exons encode the common cytoplasmic region. These neural cadherin-like cell adhesion proteins most likely play a critical role in the establishment and function of specific cell-cell connections in the brain. Alternative splicing has been described for the gamma cluster genes. Anti-Protocadherin Gamma (pan) is ideal for research in Neuroscience, Cell Adhesion and Cell Signaling.

Anti-Protocadherin Gamma (pan) Biotin Conjugated Antibody is suitable for Western Blots and Immunocytochemistry. Expect a band approximately ~100 kDa on specific lysates or tissues. It will cross react with all Gamma-protocadherins -A, -B, and -C. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: Protocadherin-Gamma-A1
Clonality: Monoclonal
Clone: S159-5
Conjugation: BAC
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty