Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRR5 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DT5
75931-348EA 518 USD InStock
75931-348
Anti-PRR5 Rabbit Polyclonal Antibody
Antibodies
Proline-rich protein 5 (PRR5), also known as Protor-1, is a 388 amino acid protein in Protor family, is thought to act as a tumor suppressor in breast and colorectal tumorigenesis. PRR5 is widely expressed and possesses two RICTOR interaction sites and a C-terminal Proline rich region. It promotes Rapamycin complex 2 (mTORC2) activity. There are four isoforms of PRR5 that are produced as a result of alternative splicing events and these isoforms play an important role in the modulation of platelet-derived growth factor signaling and in the regulation of PDGFR-beta expression.

Anti-PRR5 Antibody has been tested for use in ELISA, Western Blotting, Immunohistochemistry and Immunofluorescence. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 43 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PRR5
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty