Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PRRX1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-DT8
75931-354EA 518 USD InStock
75931-354
Anti-PRRX1 Rabbit Polyclonal Antibody
Antibodies
The paired related homeobox protein 1 (PRRX1) is a paired-type DNA-binding homeodomain that plays an important role as a key switch for neural cell lineage determination and is essential for development (1,2). PRRX1 is expressed in a number of undifferentiated mesenchymal cells in mouse embryos and adults (3). Mice with a disrupted PRRX1 gene show perinatal death with craniofacial and limb malformations (4). Low PRRX1 expression levels were significantly associated with metastasis and poor prognosis of breast and colorectal cancer (5).

Anti-PRRX1 Antibody has been tested for use in ELISA and Western Blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band at approximately 27 kDa in Western Blots of specific cell lysates and tissues.

Type: Primary
Antigen: PRRX1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty