Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-PTPH1 Rabbit Polyclonal Antibody

Supplier: Rockland Immunochemicals
600-401-X22
75931-384EA 518 USD InStock
75931-384
Anti-PTPH1 Rabbit Polyclonal Antibody
Antibodies
Protein-tyrosine Phosphatase H1 (PTPH1) has recently been identified as a specific p38γ MAPK phosphatase which is mediated through PDZ interaction. Ras has been demonstrated to increase both p38 gamma and PTPH1 protein expression, and there is a coupling of increased p38 gamma and PTPH1 protein expression in primary colon cancer tissues. Phosphorylation of PTPH1 at Ser459 leads to PTPH1 stabilization, which plays an important role in Ras oncogenesis and stress response. Additionally, phosphorylation of PTPH1 Ser459 reveals a novel mechanism by which MAPK signals through PTPH1 to regulate cellular response. PTPH1 pS459 Antibody is ideal for researchers interested in cancer research.

Anti-Phospho-Ser459 PTPH1 Antibody is stored with 100 μg per ml BSA. It is suitable for Western Blots and is specific for the ~104 kDa PTPH1 protein phosphorylated at Ser459. Immunolabeling is blocked by preadsorption Cof the antibody with the phosphopeptide used as antigen but not by the corresponding dephosphopeptide. Immunolabeling is also completely eliminated by treatment with lambda-phosphatase. Specific conditions for reactivity should be optimized by the end user.

Type: Primary
Antigen: PTPH1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty